Learn More
PDIA3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PDIA3.
Supplier: Abnova Corporation PAB6295
Description
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates. [provided by RefSeq
Sequence: C-EEKPKKKKKAQEDLSpecifications
PDIA3 | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
PDIA3 | |
PDIA3 | |
A synthetic peptide corresponding to human PDIA3. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PDIA3. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
ER60/ERp57/ERp60/ERp61/GRP57/GRP58/HsT17083/P58/PI-PLC | |
Goat | |
Antigen affinity purification | |
RUO | |
2923 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only