Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peg12, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Peg12.
Supplier: Abnova Corporation PAB6137
Description
Sequence: C-SLAANNPGGHSTNLSpecifications
Peg12 | |
Polyclonal | |
Unconjugated | |
ELISA (1:2000) The optimal working dilution should be determined by the end user. | |
Peg12 | |
Peg12 | |
A synthetic peptide corresponding to C-terminus of mouse Peg12. | |
100 μg | |
Primary | |
Mouse | |
Liquid |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of Peg12. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
Frat3 | |
Goat | |
Antigen affinity purification | |
RUO | |
27412 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction