Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPLA3, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133396 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-396 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-396 Supplier Abnova Corporation Supplier No. PAB7591
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of PNPLA3.

The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes. [provided by RefSeq

Sequence: EHDICPKVKSTN

Specifications

Antigen PNPLA3
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of PNPLA3.
Dilution ELISA (1:128000) Western Blot (0.05-0.2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(4-6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene PNPLA3
Gene Alias ADPN/C22orf20/iPLA(2)epsilon
Gene Symbols PNPLA3
Host Species Goat
Immunogen A synthetic peptide corresponding to internal region of human PNPLA3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80339
Test Specificity This antibody is raised against a peptide derived from the mouse protein sequence and this peptide differs from human only in position 2.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.