Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLK, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of POLK.
Supplier: Abnova Corporation PAB11246
Specifications
POLK | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of POLK. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
DINB1/DINP/POLQ | |
Goat | |
100 μg | |
Primary | |
Expect reactivity with multiple splice variants of this protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000-1:10000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
POLK | |
POLK | |
A synthetic peptide corresponding to amino acids 817-830 of human POLK. | |
RUO | |
51426 | |
Chimpanzee, Human, Mouse, Orangutan | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction