Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLL, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132485 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-485 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-485 Supplier Abnova Corporation Supplier No. PAB6543
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of POLL.

Sequence: C-LGLPYREPAERDW

Specifications

Antigen POLL
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of POLL.
Dilution ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene POLL
Gene Alias BETA-N/FLJ46002/POL-KAPPA
Gene Symbols POLL
Host Species Goat
Immunogen A synthetic peptide corresponding to human POLL.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27343
Test Specificity This antibody may crossreact with Pol beta (NP_002681), an evolutionarilly related polymerase with a predicted size of 39 KDa. The overall % of identity is not very high (about 32% in the core) but considering only the that we used as immunogen the two share the sequence YREP**R** at the C-terminus. Further the protein structures appear to be very similar in this region.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.