Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU3F3, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89120137 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-137 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-137 Supplier Abnova Corporation Supplier No. PAB18580
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of POU3F3.

POU3F3 is a member of the class III POU family of transcription factors (see POU3F1; MIM 602479) that are expressed in the central nervous system. The POU domain in these proteins is required for high affinity binding to octamer DNA sequences Sumiyama et al. (1996) [PubMed 8703082].[supplied by OMIM

Sequence: C-HMLSHAHQWVTAL

Specifications

Antigen POU3F3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of POU3F3.
Dilution ELISA (1:4000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene POU3F3
Gene Alias BRN1/OTF8
Gene Symbols POU3F3
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human POU3F3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5455
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.