Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU4F3, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89120558 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-558 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-558 Supplier Abnova Corporation Supplier No. PAB19005
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of POU4F3.

This gene encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of nonsyndromic sensorineural deafness autosomal dominant type 15. [provided by RefSeq

Sequence: C-EAAYREKNSKPE

Specifications

Antigen POU4F3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of POU4F3.
Dilution ELISA (1:8000) Western Blot (0.2-0.6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene POU4F3
Gene Alias BRN3C/DFNA15/MGC138412
Gene Symbols POU4F3
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human POU4F3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5459
Test Specificity This antibody is expected not to cross-react to the similar POU4F1 and POU4F2.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.