Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R3A Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-397 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-397 Supplier Abnova Corporation Supplier No. PAB7592
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of PPP2R3A.

Protein phosphatase 2 (formerly named type 2A) is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B″/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B″ family. The B″ family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B″. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: C-QKDVENDGPEPSD

Specifications

Antigen PPP2R3A
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of PPP2R3A.
Dilution ELISA (1:16000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene PPP2R3A
Gene Alias PPP2R3/PR130/PR72
Gene Symbols PPP2R3A
Host Species Goat
Immunogen A synthetic peptide corresponding to human PPP2R3A.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5523
Test Specificity This antibody is expected to recognize both reported isoforms (NP_002709.2; NP_871626.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.