Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-PRNP, Polyclonal, Abnova™

Catalog No. 89116887 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-887 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-887 Supplier Abnova Corporation Supplier No. PAB14028

Specifications

Antigen PRNP
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In buffer containing 0.09% sodium azide
Gene Symbols PRNP
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 79-97 of human PRNP.
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) 5621
Target Species Bovine, Hamster, Human, Sheep
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.