Learn More
PYCARD, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PYCARD.
Supplier: Abnova Corporation PAB6098
Description
This gene encodes an adaptor protein that is composed of two protein-protein interaction domains: a N-terminal PYRIN-PAAD-DAPIN domain (PYD) and a C-terminal caspase-recruitment domain (CARD). The PYD and CARD domains are members of the six-helix bundle death domain-fold superfamily that mediates assembly of large signaling complexes in the inflammatory and apoptotic signaling pathways via the activation of caspase. In normal cells, this protein is localized to the cytoplasm; however, in cells undergoing apoptosis, it forms ball-like aggregates near the nuclear periphery. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-RESQSYLVEDLERSSpecifications
PYCARD | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (1-3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
PYCARD | |
PYCARD | |
A synthetic peptide corresponding to C-terminus of human PYCARD. | |
100 μg | |
Primary | |
This antibody is expected to recognize all three reported human isoforms according to NP_037390.2, NP_660183.1 and NP_660184.1. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PYCARD. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
ASC/CARD5/MGC10332/TMS/TMS-1/TMS1 | |
Goat | |
Antigen affinity purification | |
RUO | |
29108 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only