Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Shandon™ Iris Curved Scissors, Curved, premium, 4.5 in. (11.4cm)
Epredia™ Shandon Iris Curved Scissors are available in Premium and Standard grades.
Supplier: Epredia™ 31212
Description
Consistent Quality
- Properly hardened, tempered and passivated
- Patterns, dimensions and finish held to exacting standards
- Guaranteed to be free from defects in material and workmanship
Specifications
Iris Scissors | |
Standard | |
11.4 cm |
Curved | |
4.5 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction