Learn More
RAD23B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RAD23B.
Supplier: Abnova Corporation PAB10058
Description
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in the nucleotide excision repair (NER). This protein was found to be a component of the protein complex that specifically complements the NER defect of xeroderma pigmentosum group C (XP-c) cell extracts in vitro. This protein was also shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, and thus this protein may be involved in the ubiquitin mediated proteolytic pathway in cells. [provided by RefSeq
Specifications
RAD23B | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of RAD23B. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
HHR23B/HR23B/P58 | |
Goat | |
100 μg | |
Primary | |
Canine, Chimpanzee, Human, Mouse, Rat, Yeast | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:4000-1:16000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
RAD23B | |
RAD23B | |
A synthetic peptide corresponding to amino acids 163-176 of human RAD23B. | |
RUO | |
5887 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only