Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137689 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-689 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-689 Supplier Abnova Corporation Supplier No. PAB13816
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native Respiratory Syncytial Virus.

Specifications

Antigen Respiratory Syncytial Virus
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native Respiratory Syncytial Virus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Goat
Immunogen Native purified of human respiratory syncytial virus.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity All RSV viral antigens including RSV-A and RSV-B. Reacts well with bovine isolates. Does not react with Para 1-3, Influenza A & B or Adenovirus by IFA. Does not react with HEp-2 cells and WI-38 cells.
Target Species Respiratory Syncytial virus (RSV)
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.