Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF125, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132555 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-555 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-555 Supplier Abnova Corporation Supplier No. PAB6638
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of RNF125.

This gene encodes a novel E3 ubiquitin ligase that contains an N-terminal RING finger domain. The encoded protein may function as a positive regulator in the T-cell receptor signaling pathway. [provided by RefSeq

Sequence: C-RSLLEYVNHSNTT

Specifications

Antigen RNF125
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of RNF125.
Dilution ELISA (1:8000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene RNF125
Gene Alias FLJ20456/MGC21737/TRAC1
Gene Symbols RNF125
Host Species Goat
Immunogen A synthetic peptide corresponding to human RNF125.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54941
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.