Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF2 Goat anti-Bovine, Chicken, Chimpanzee, Dog, Frog, Human, Mouse, Orangutan, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114711 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-711 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-711 Supplier Abnova Corporation Supplier No. PAB10232
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of RNF2.

Polycomb group (PcG) of proteins form the multiprotein complexes that are important for the transcription repression of various genes involved in development and cell proliferation. The protein encoded by this gene is one of the PcG proteins. It has been shown to interact with, and suppress the activity of, transcription factor CP2 (TFCP2/CP2). Studies of the mouse counterpart suggested the involvement of this gene in the specification of anterior-posterior axis, as well as in cell proliferation in early development. This protein was also found to interact with huntingtin interacting protein 2 (HIP2), an ubiquitin-conjugating enzyme, and possess ubiquitin ligase activity. [provided by RefSeq

Specifications

Antigen RNF2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description goat polyclonal antibody raised against synthetic peptide of RNF2.
Dilution ELISA (1:5000-1:25000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene RNF2
Gene Alias BAP-1/BAP1/DING/HIPI3/RING1B/RING2
Gene Symbols RNF2
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 189-201 of human RNF2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6045
Test Specificity This affinity purified antibody is directed against human RING1B protein.
Target Species Bovine, Canine, Chicken, Chimpanzee, Frog, Human, Mouse, Orangutan, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.