Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF34 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89132353 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-353 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-353 Supplier Abnova Corporation Supplier No. PAB6360
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of RNF34.

The protein encoded by this gene contains a RINF finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein interacts with DNAJA3/hTid-1, which is a DnaJ protein reported to function as a modulator of apoptosis. Overexpression of this gene in Hela cells was shown to confer the resistance to TNF-alpha induced apoptosis, suggesting an anti-apoptotic function of this protein. This protein can be cleaved by caspase-3 during the induction of apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq

Sequence: KAGATSMWASCC

Specifications

Antigen RNF34
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of RNF34.
Dilution ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene RNF34
Gene Alias FLJ21786/RFI/RIF/RIFF
Gene Symbols RNF34
Host Species Goat
Immunogen A synthetic peptide corresponding to N-terminus of human RNF34.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80196
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.