Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SALL1 Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133106 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-106 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-106 Supplier Abnova Corporation Supplier No. PAB7277
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SALL1.

The protein encoded by this gene is a zinc finger transcriptional repressor and may be part of the NuRD histone deacetylase complex (HDAC). Defects in this gene are a cause of Townes-Brocks syndrome (TBS) as well as bronchio-oto-renal syndrome (BOR). Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-QDSKDTPTSHVPS

Specifications

Antigen SALL1
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SALL1.
Dilution ELISA (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene SALL1
Gene Alias HSAL1/TBS/ZNF794
Gene Symbols SALL1
Host Species Goat
Immunogen A synthetic peptide corresponding to human SALL1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6299
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.