Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMSN1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132587 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-587 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-587 Supplier Abnova Corporation Supplier No. PAB6673
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SAMSN1.

SAMSN1 is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM (sterile alpha motif) domains (Claudio et al., 2001 [PubMed 11536050]).[supplied by OMIM

Sequence: IRLDTLSASLGRSC

Specifications

Antigen SAMSN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SAMSN1.
Dilution ELISA (1:64000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene SAMSN1
Gene Alias HACS1/NASH1/SASH2/SH3D6B
Gene Symbols SAMSN1
Host Species Goat
Immunogen A synthetic peptide corresponding to human SAMSN1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64092
Test Specificity The immunogen sequence is present in NP_071419.2 (and in AAK07746) but is no longer present in the latest reference sequence (NP_071419.3 at 05/10/05). This latest version has not been subject to final review by NCBI. This antibody may be recognising one particular isoform.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.