Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAR1B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SAR1B.
Supplier: Abnova Corporation PAB7320
Description
Sequence: C-DHERLLESKEELDSpecifications
SAR1B | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) Western Blot (0.03-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
SAR1B | |
SAR1B | |
A synthetic peptide corresponding to human SAR1B. | |
100 μg | |
Primary | |
Both reported variants (NP_001028675.1 and NP_057187.1) represent identical protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SAR1B. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
CMRD/GTBPB/SARA2 | |
Goat | |
Antigen affinity purification | |
RUO | |
51128 | |
Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction