Learn More
Description
- Whole antiserum
- Neat serum with 0.09% Sodium Azide
- Applications:
- Western Blot: Recommended use at 1:100 to 1:1000 dilution (dilution may vary depending on amount of antigen and sensitivity of assay).
- ELISA: Recommended use at 1:1000 to 1:10,000 dilution.
Specifications
Specifications
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Host Species | Goat |
| Immunogen | Detergent disrupted SARS-CoV-2 purified from Vero cells |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | SARS-CoV-2 |
| Content And Storage | Store at -10°C or below. Repetitive freezing and thawing are not recommended (aliquot as necessary). Thawed material may be stored at 4°C for short-term usage. |
| Product Type | Primary Antibody Serum |
| Show More |
RUO only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
