Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SET, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132094 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-094 Supplier Abnova Corporation Supplier No. PAB6048
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SET.

Sequence: SAPAAKVSKKELNS-C

Specifications

Antigen SET
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SET.
Dilution ELISA (1:1000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene SET
Gene Alias 2PP2A/I2PP2A/IGAAD/IPP2A2/PHAPII/TAF-I/TAF-IBETA
Gene Symbols SET
Host Species Goat
Immunogen A synthetic peptide corresponding to human SET.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6418
Test Specificity The peptide sequence used (from AAC50460.1) differs by one residue from the RefSeq entry (NP_003002.1) and the three other Genbank protein sequences referenced in the Locuslink entry. It is not known whether this difference is due to a sequencing error or polymophism but this antibody is expected to recognize both sequences.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.