Learn More
SIRT3 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SIRT3.
Supplier: Abnova Corporation PAB6891
Description
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family. Two alternatively spliced transcript variants that encode different proteins have been described for this gene. [provided by RefSeq
Sequence: C-QRETGKLDGPDKSpecifications
SIRT3 | |
Polyclonal | |
Unconjugated | |
ELISA (1:128000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
SIRT3 | |
SIRT3 | |
A synthetic peptide corresponding to human SIRT3. | |
100 μg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_036371.1 and NP_001017524.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SIRT3. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
SIR2L3 | |
Goat | |
Antigen affinity purification | |
RUO | |
23410 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only