Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC24A5, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132895 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-895 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-895 Supplier Abnova Corporation Supplier No. PAB7036
Only null left
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SLC24A5.

This gene is a member of the potassium-dependent sodium/calcium exchanger family and encodes an intracellular membrane protein with 2 large hydrophilic loops and 2 sets of multiple transmembrane-spanning segments. Sequence variation in this gene has been associated with differences in skin pigmentation. [provided by RefSeq

Sequence: TTPDCRKKFWKN

Specifications

Antigen SLC24A5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SLC24A5.
Dilution ELISA (1:2000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene SLC24A5
Gene Alias JSX/NCKX5/SHEP4
Gene Symbols SLC24A5
Host Species Goat
Immunogen A synthetic peptide corresponding to human SLC24A5.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283652
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.