Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC46A1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89121225 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-225 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-225 Supplier Abnova Corporation Supplier No. PAB19698
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SLC46A1.

This gene encodes a transmembrane proton-coupled folate transporter protein that facilitates the movement of folate and antifolate substrates across cell membranes optimally in acidic pH environments. This protein is also expressed in the brain and choroid plexus where it transports folates into the central nervous system. This protein further functions as a transmembrane heme transporter in duodenal enterocytes and, potentially, in other tissues like liver and kidney. Its localization to the apical membrane or cytoplasm of intestinal cells is modulated by dietary iron levels. Mutations in this gene cause the autosomal recessive hereditary folate malabsorption (HFM) disease. HFM is characterized by folate deficiency due to reduced intestinal folate absorption and subsequent anemia, hypoimmunoglobulinemia, and recurrent infections. [provided by RefSeq

Sequence: C-LKEPKSTRLFTFRH

Specifications

Antigen SLC46A1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SLC46A1.
Dilution Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene SLC46A1
Gene Alias HCP1/MGC9564/PCFT
Gene Symbols SLC46A1
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 233-247 at internal region of human SLC46A1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113235
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.