Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A12, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SLC6A12.
Supplier: Abnova Corporation PAB27630
Description
Sequence: CLDGSAGRNFGPSPTSpecifications
SLC6A12 | |
Polyclonal | |
Unconjugated | |
ELISA (1:128000) Western Blot (1-3ug/ml) The optimal working dilution should be determined by the end user. | |
SLC6A12 | |
SLC6A12 | |
A synthetic peptide corresponding to C-terminus of human SLC6A12. | |
100 μg | |
Primary | |
Human |
Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SLC6A12. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
BGT-1/BGT1/FLJ38727 | |
Goat | |
Affinity Purified | |
RUO | |
6539 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction