Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A3, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89126317 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-317 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-317 Supplier Abnova Corporation Supplier No. PAB24600
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SLC6A3.

The dopamine transporter (DAT1) mediates the active reuptake of dopamine from the synapse and is a principal regulator of dopaminergic neurotransmission. The DAT1 gene has been implicated in human disorders such as parkinsonism, Tourette syndrome, and substance abuse (Vandenbergh et al., 1992 [PubMed 1359373]).[supplied by OMIM

Sequence: C-QLTSSTLTNPRQSP

Specifications

Antigen SLC6A3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SLC6A3.
Dilution ELISA (1:4000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene SLC6A3
Gene Alias DAT/DAT1
Gene Symbols SLC6A3
Host Species Goat
Immunogen A synthetic peptide corresponding to near N-terminus of human SLC6A3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6531
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.