Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC9A3R2 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89132772 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-772 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-772 Supplier Abnova Corporation Supplier No. PAB6889
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SLC9A3R2.

NHE3 (SLC9A3; MIM 182307) is a sodium/hydrogen exchanger in the brush border membrane of the proximal tubule, small intestine, and colon that plays a major role in transepithelial sodium absorption. SLC9A3R2, as well as SLC9A3R1 (MIM 604990) and protein kinase A phosphorylation, may play a role in NHE3 regulation.[supplied by OMIM

Sequence: C-DGSAWKQDPFQ

Specifications

Antigen SLC9A3R2
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SLC9A3R2.
Dilution ELISA (1:32000) Western blot (0.3-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene SLC9A3R2
Gene Alias E3KARP/MGC104639/NHE3RF2/NHERF-2/NHERF2/OCTS2/SIP-1/SIP1/TKA-1
Gene Symbols SLC9A3R2
Host Species Goat
Immunogen A synthetic peptide corresponding to human SLC9A3R2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9351
Test Specificity This variant has an extra insertion of 11 residues compared to NP_004776.3. The immunizing peptide represents this insertion.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.