Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMC5, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SMC5.
Supplier: Abnova Corporation PAB17817
Description
Sequence: C-TSKHLEHKDNELRQSpecifications
SMC5 | |
Polyclonal | |
Unconjugated | |
ELISA (1:128000) The optimal working dilution should be determined by the end user. | |
SMC5 | |
SMC5 | |
A synthetic peptide corresponding to amino acids at internal region of human SMC5. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SMC5. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
KIAA0594/SMC5L1 | |
Goat | |
Antigen affinity purification | |
RUO | |
23137 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction