Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX26 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89132365 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-365 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-365 Supplier Abnova Corporation Supplier No. PAB6375
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SNX26.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. The specific function of this protein has not been elucidated. Alternative splice variants have been described but their full-length nature has not been determined. [provided by RefSeq

Sequence: SWSLHSEGQTRSYC

Specifications

Antigen SNX26
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SNX26.
Dilution ELISA (1:8000) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene SNX26
Gene Alias FLJ39019/TCGAP
Gene Symbols SNX26
Host Species Goat
Immunogen A synthetic peptide corresponding to human SNX26.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115703
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.