Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPINT2 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SPINT2.
Supplier: Abnova Corporation PAB19048
Description
Sequence: C-PSAPRRQDSEDHSpecifications
SPINT2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
SPINT2 | |
SPINT2 | |
A synthetic peptide corresponding to amino acids at internal region of human SPINT2. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SPINT2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
FLJ45571/HAI-2/HAI2/Kop/PB | |
Goat | |
Antigen affinity purification | |
RUO | |
10653 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction