Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPINT2 Goat anti-Chicken, Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89129124 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-124 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-124 Supplier Abnova Corporation Supplier No. PAB27614
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of SPINT2.

Sequence: SGDDKEQLVKNT

Specifications

Antigen SPINT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of SPINT2.
Dilution ELISA (1:1000) Western Blot (1-3ug/ml) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene SPINT2
Gene Alias FLJ45571/HAI-2/HAI2/Kop/PB
Gene Symbols SPINT2
Host Species Goat
Immunogen A synthetic peptide corresponding to human SPINT2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10653
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.