Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBL1X/TBL1XR1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132153 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-153 Supplier Abnova Corporation Supplier No. PAB6110
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of TBL1X/TBL1XR1.

Sequence: SASDGSVCVLDLRK

Specifications

Antigen TBL1X/TBL1XR1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of TBL1X/TBL1XR1.
Dilution ELISA (1:4000) Western blot (3-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene Alias EBI/SMAP55/TBL1
Gene Symbols TBL1X
Host Species Goat
Immunogen A synthetic peptide corresponding to human TBL1X, TBL1XR1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6907/79718
Test Specificity This antibody is also expected to recognize the human protein TBLR1 (NP_078941), which is very similar.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.