Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFB1M, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89118493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-493 Supplier Abnova Corporation Supplier No. PAB16639
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of TFB1M.

The transcription of genes from mitochondrial DNA requires a mitochondrial RNA polymerase (see POLRMT, MIM 601778) and a DNA-binding transcription factor (see TFAM, MIM 600438). Transcription factor B1 (TFB1M) is a part of this transcription complex.[supplied by OMIM

Sequence: C-ELKRRKSKNEEKE

Specifications

Antigen TFB1M
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of TFB1M.
Dilution ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene TFB1M
Gene Alias CGI-75/CGI75/mtTFB/mtTFB1
Gene Symbols TFB1M
Host Species Goat
Immunogen A synthetic peptide corresponding to human TFB1M.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51106
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.