Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBR3 Goat, Polyclonal Antibody, Abnova™

Catalog No. 89115206 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-206 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-206 Supplier Abnova Corporation Supplier No. PAB11592
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of TGFBR3.

Transforming growth factor (TGF)-beta is a multifunctional cytokine that modulates several tissue development and repair processes, including cell differentiation, cell cycle progression, cellular migration, adhesion, and extracellular matrix production. Three TGF-beta forms are encoded by separate genes: TGFB1 (MIM 190180), TGFB2 (MIM 190220), and TGFB3 (MIM 190230). The diverse effects of TGF-beta are mediated by the TGF-beta receptors and cell surface-binding proteins. Three TGF-beta receptors exist: type I (TGFBR1; MIM 190181), type II (TFGBR2; MIM 190182), and type III (TGFBR3). TGFBR3 is a glycoprotein that binds TGFB and exists in both a membrane-bound and a soluble form (Johnson et al., 1995 [PubMed 8530052]).[supplied by OMIM

Sequence: C-TKSIRDDIPSTQGN

Specifications

Antigen TGFBR3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of TGFBR3.
Dilution ELISA (1:32000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene TGFBR3
Gene Alias BGCAN/betaglycan
Gene Symbols TGFBR3
Host Species Goat
Immunogen A synthetic peptide corresponding to human TGFBR3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7049
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.