Learn More
TMPRSS4 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of TMPRSS4.
Supplier: Abnova Corporation PAB15676
Description
This gene encodes a member of the serine protease family. Serine proteases are known to be involved in a variety of biological processes, whose malfunction often leads to human diseases and disorders. This gene was identified as a gene overexpressed in pancreatic carcinoma. The encoded protein is membrane bound with a N-terminal anchor sequence and a glycosylated extracellular region containing the serine protease domain. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: HCFRKHTDVFNWKSpecifications
TMPRSS4 | |
Polyclonal | |
Unconjugated | |
ELISA (1:1000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
TMPRSS4 | |
TMPRSS4 | |
A synthetic peptide corresponding to amino acids 245-57 of human TMPRSS4. | |
100 μg | |
Primary | |
Approximately 48 KDa band observed in lysates of cell lines HEK293 and A-549 (calculated MW of 48.2 KDa according to NP_063947.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised aganist synthetic peptide of TMPRSS4. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
MT-SP2/TMPRSS3 | |
Goat | |
Antigen affinity purification | |
RUO | |
56649 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only