Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNKS, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117738 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-738 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-738 Supplier Abnova Corporation Supplier No. PAB15673
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist partial recombinant TNKS.

Sequence: RECOMBINANT

Specifications

Antigen TNKS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist partial recombinant TNKS.
Dilution Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene TNKS
Gene Alias PARP-5a/PARP5A/PARPL/TIN1/TINF1/TNKS1
Gene Symbols TNKS
Host Species Goat
Immunogen Recombinant protein corresponding to N-terminus 120 amino acids of human TNKS.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8658
Test Specificity In transfected HEK293 transiently expressing TANK1 a band of approximately 142 KDa is observed. This band is not observed in the non-transfected HEK293. No cross-reactivity expected to TANK2.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.