Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNKS2 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117737 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-737 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-737 Supplier Abnova Corporation Supplier No. PAB15672
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist partial recombinant TNKS2.

Sequence: RECOMBINANT2

Specifications

Antigen TNKS2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist partial recombinant TNKS2.
Dilution Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene TNKS2
Gene Alias PARP-5b/PARP-5c/PARP5B/PARP5C/TANK2/TNKL
Gene Symbols TNKS2
Host Species Goat
Immunogen Recombinant protein corresponding to 73 amino acids from 800-900 region of human TNKS2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80351
Test Specificity In transfected HEK293 transiently expressing TANK2 a band of approximately 127 KDa is observed. This band is not observed in the non-transfected HEK293. No cross-reactivity expected with TANK1.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.