missing translation for 'onlineSavingsMsg'
Learn More

TPD52L3, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133183 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-183 Supplier Abnova Corporation Supplier No. PAB7362
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of TPD52L3.

This gene encodes a member of the tumor protein D52-like family of proteins. These proteins are characterized by an N-terminal coiled-coil motif that is used to form homo- and heteromeric complexes with other tumor protein D52-like proteins. The encoded protein may play a role in spermatogenesis. Alternate splicing results in multiple transcript variants. [provided by RefSeq

Sequence: PHARTETSVGTYESH-C

Specifications

Antigen TPD52L3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of TPD52L3.
Dilution ELISA (1:4000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene TPD52L3
Gene Alias MGC26757/MGC45374/NYD-SP25/hD55
Gene Symbols TPD52L3
Host Species Goat
Immunogen A synthetic peptide corresponding to human TPD52L3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89882
Test Specificity This antibody is expected to recognize all three reported isoforms (NP_277051.3; NP_001001874.1; NP_001001875.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.