Learn More
TRIM69, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TRIM69.
Supplier: Abnova Corporation PAB6364
Description
This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Sequence: C-GENKEPLHILHPQSpecifications
TRIM69 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5-3.8 ug/mL) The optimal working dilution should be determined by the end user. | |
TRIM69 | |
TRIM69 | |
A synthetic peptide corresponding to C-terminus of human TRIM69. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of TRIM69. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
HSD34/RNF36/Trif | |
Goat | |
Antigen affinity purification | |
RUO | |
140691 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only