Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I-Cardiac Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89137709 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-709 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-709 Supplier Abnova Corporation Supplier No. PAB13849
Add to Cart
Add to Cart

Goat polyclonal antibody raised against Troponin I-Cardiac.

Specifications

Antigen Troponin I-Cardiac
Applications ELISA, Immunoblot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against Troponin I-Cardiac.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM sodium phosphate buffer, pH 7.2 (0.85% NaCl, 0.05% sodium azide)
Host Species Goat
Immunogen Purified human cardiac Troponin I.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for human Cardiac Troponin I. Recognizes epitopes located in the 27-39 region of Troponin I. Cross reactivity with human skeletal Troponin I is 0.02% by ELISA.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.