Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tspo, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117721 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-721 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-721 Supplier Abnova Corporation Supplier No. PAB15654
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist synthetic peptide of Tspo.

Sequence: C-RDNSGRRGGSRLPE

Specifications

Antigen Tspo
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist synthetic peptide of Tspo.
Dilution ELISA (1:8000) Western Blot (0.2-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene Tspo
Gene Alias Bzrp/IBP/PBR
Gene Symbols Tspo
Host Species Goat
Immunogen A synthetic peptide corresponding to mouse Tspo.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12257
Test Specificity Approximately 18 KDa band observed in mouse spleen lysates (calculated MW of 18.8 KDa according to NP_033905.3).
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.