Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUSC2, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117743 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-743 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-743 Supplier Abnova Corporation Supplier No. PAB15678
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist synthetic peptide of TUSC2.

This gene is a highly conserved lung cancer candidate gene. No other information about this gene is currently available. [provided by RefSeq

Sequence: C-TKNGQKRAKLRRVH

Specifications

Antigen TUSC2
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist synthetic peptide of TUSC2.
Dilution ELISA (1:2000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene TUSC2
Gene Alias C3orf11/FUS1/PAP/PDAP2
Gene Symbols TUSC2
Host Species Goat
Immunogen A synthetic peptide corresponding to interal region of human TUSC2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11334
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.