Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC5 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121194 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-194 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-194 Supplier Abnova Corporation Supplier No. PAB19669
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of TXNDC5.

This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-RDLESLREYVESQLQ

Specifications

Antigen TXNDC5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of TXNDC5.
Dilution Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene TXNDC5
Gene Alias ERP46/EndoPDI/Hcc-2/MGC3178/UNQ364
Gene Symbols TXNDC5
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 281-295 at internal region of human TXNDC5.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81567
Test Specificity This antibody is expected to recognize both reported isoforms (NP_110437.2; NP_001139021.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.