Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-UCHL1, Polyclonal, Abnova™

Catalog No. 89116858 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-858 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-858 Supplier Abnova Corporation Supplier No. PAB13854

Specifications

Antigen UCHL1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In buffer containing 0.09% sodium azide
Gene Symbols UCHL1
Host Species Goat
Immunogen A synthetic peptide corresponding to human UCHL1.
Quantity 1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7345
Target Species Human
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.