Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT8, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89129083 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-083 Supplier Abnova Corporation Supplier No. PAB27572
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of UGT8.

Galactocerebrosides are abundant sphingolipids of the myelin membrane of the central nervous system and peripheral nervous system and are also present in small amounts in kidney. The key enzymatic step in the biosynthesis of galactocerebrosides consists of the transfer of galactose to ceramide catalyzed by UDP-galactose ceramide galactosyltransferase (CGT, EC 2.4.1.45). The enzyme encoded by the CGT gene is the first involved in complex lipid biosynthesis in the myelinating oligodendrocyte.[supplied by OMIM

Sequence: NGKYKRNGHIK

Specifications

Antigen UGT8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of UGT8.
Dilution ELISA (1:64000) Western Blot (1-3ug/ml) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene UGT8
Gene Alias CGT
Gene Symbols UGT8
Host Species Goat
Immunogen A synthetic peptide corresponding to internal region of human UGT8.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7368
Target Species Canine, Chicken, Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.