Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VANGL2, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133045 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-045 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-045 Supplier Abnova Corporation Supplier No. PAB7207
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of VANGL2.

The VANGL2 gene encodes a homolog of Drosophila 'strabismus/Van Gogh' (Stbm/Vang), a component of the frizzled-dishevelled tissue polarity pathway (Kibar et al., 2001 [PubMed 11431695]).[supplied by OMIM

Sequence: C-QKYLRTTKQQPYH

Specifications

Antigen VANGL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of VANGL2.
Dilution ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene VANGL2
Gene Alias KIAA1215/LPP1/LTAP/MGC119403/MGC119404/STB1/STBM/STBM1
Gene Symbols VANGL2
Host Species Goat
Immunogen A synthetic peptide corresponding to human VANGL2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57216
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.