Learn More
VPS29 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of VPS29.
Supplier: Abnova Corporation PAB6574
Description
This gene belongs to a group of vacuolar protein sorting (VPS) genes that, when functionally impaired, disrupt the efficient delivery of vacuolar hydrolases. The protein encoded by this gene is a component of a large multimeric complex, termed the retromer complex, which is involved in retrograde transport of proteins from endosomes to the trans-Golgi network. This VPS protein may be involved in the formation of the inner shell of the retromer coat for retrograde vesicles leaving the prevacuolar compartment. Alternative splice variants encoding different isoforms, and usage of multiple polyadenylation sites have been found for this gene. [provided by RefSeq
Sequence: C-DDVKVERIEYKKPSpecifications
VPS29 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
VPS29 | |
VPS29 | |
A synthetic peptide corresponding to human VPS29. | |
100 μg | |
Primary | |
This antibody is expected to recognize both human isoforms (represented by NP_057310.1; NP_476528.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of VPS29. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
DC15/DC7/DKFZp564F0223/FLJ20492/PEP11 | |
Goat | |
Antigen affinity purification | |
RUO | |
51699 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only