Learn More
WT1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of WT1.
Supplier: Abnova Corporation PAB7070
Description
This gene encodes a transcription factor that contains four zinc-finger motifs at the C-terminus and a proline/glutamine-rich DNA-binding domain at the N-terminus. It has an essential role in the normal development of the urogenital system, and it is mutated in a small subset of patients with Wilm's tumors. Multiple transcript variants, resulting from alternative splicing at two coding exons, have been well characterized. There is also evidence for the use of non-AUG (CUG) translation initiation site upstream of, and in-frame with the first AUG, leading to additional isoforms. Authors of PMID:7926762 also provide evidence that WT1 mRNA undergoes RNA editing in human and rat, and that this process is tissue-restricted and developmentally regulated. [provided by RefSeq]
Sequence: QDPASTCVPEPASQHSpecifications
| WT1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| WT1 | |
| WT1 | |
| A synthetic peptide corresponding to human WT1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize all four reported isoforms (NP_000369.3, NP_077742.2, NP_077743.2, NP_077744.3). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of WT1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| GUD/WAGR/WIT-2/WT33 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 7490 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only