Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP112, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117749 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-749 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-749 Supplier Abnova Corporation Supplier No. PAB15684
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist synthetic peptide of ZFP112.

Sequence: C-EREDDIENSHLKSYQ

Specifications

Antigen ZFP112
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist synthetic peptide of ZFP112.
Dilution ELISA (1:4000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene ZFP112
Gene Alias ZNF228
Gene Symbols ZFP112
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human ZFP112.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7771
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.