Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat F(ab')2 anti-Mouse IgG (H+L) Secondary Antibody, Rhodamine (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NB1205867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205867 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205867 Supplier Novus Biologicals Supplier No. NB1205867
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Rhodamine
Dilution Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Host Species Goat
Immunogen Mouse IgG whole molecule
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity No reaction was observed against anti-Pepsin, anti-Goat IgG F(c) or Bovine, Horse, Human, Rabbit, Rat and Sheep Serum Proteins.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.